ε-Caprolactam 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Azepan-2-one, 2-Oxohexamethyleneimine, 6-Aminocaproic acid lactam

10823-0010 10823-0025 10823-1000 10823-2500
TS10823-0010EA 48 USD
TS10823-0010 TS10823-0025 TS10823-1000 TS10823-2500
ε-Caprolactam 99+%
ε-Caprolactam
Formula: C₆H₁₁NO
MW: 113.16 g/mol
Boiling Pt: 268 °C (760 mmHg)
Melting Pt: 68…71 °C
Flash Pt: 152 °C
Storage Temperature: Ambient
MDL Number: MFCD00006936
CAS Number: 105-60-2
EINECS: 203-313-2

Order Now

Specification Test Results

Appearance White Crystals or flakes
Infrared spectrum Conforms
Melting point 68 to 71 °C
GC ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR