3-Sulfolene 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-sulpholene, Butadiene sulphone

10761-0025 10761-1000 10761-5000
TS10761-0025EA 265 USD
TS10761-0025 TS10761-1000 TS10761-5000
3-Sulfolene 98%
3-Sulpholene
Formula: C₄H₆O₂S
MW: 118.16 g/mol
Melting Pt: 63…66 °C
Flash Pt: 112 °C
Storage Temperature: Ambient
MDL Number: MFCD00005481
CAS Number: 77-79-2
EINECS: 201-059-7
UN: 3335
ADR: 9,III

Order Now

Specification Test Results

Appearance White to off-white Adhering powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 63 to 67 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR