2-Bromopyridine 99%

Supplier: Thermo Scientific Chemicals
Danger

10740-0250 10740-1000 10740-5000
TS10740-0250EA 59 USD
TS10740-0250 TS10740-1000 TS10740-5000
2-Bromopyridine 99%
2-Bromopyridine
Formula: C₅H₄BrN
MW: 158 g/mol
Boiling Pt: 192…194 °C (761 mmHg)
Density: 1.65 g/cm³
Flash Pt: 54 °C
Storage Temperature: Ambient
MDL Number: MFCD00006219
CAS Number: 109-04-6
EINECS: 203-641-6
UN: 2929
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.5700 to 1.5730 (20°C, 589 nm)
GC ≥98.5 %
Color scale ≤20 APHA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR