1-Benzyl-4-piperidone 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N-Benzyl-4-piperidone, 1-Benzylpiperidin-4-one, 1-Benzyl-4-piperidinone

10613-1000 10613-2500
TS10613-1000EA 98 USD
TS10613-1000 TS10613-2500
1-Benzyl-4-piperidone 99%
1-Benzyl-4-piperidone
Formula: C₁₂H₁₅NO
MW: 189.26 g/mol
Boiling Pt: 133…135 °C (7 mmHg)
Density: 1.06 g/cm³
Flash Pt: 71 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006192
CAS Number: 3612-20-2
EINECS: 222-782-4

Order Now

Specification Test Results

Appearance Clear colorless to straw yellow Viscous liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5390 to 1.5420 (20°C, 589 nm)
Water ≤0.5 %
GC ≥98.5 %
Titration with HClO4 98 to 102 %
UV at 400 nm A: ≤0.01
Color scale ≤10 APHA
Stabilizer 50 to 100 ppm Amylene
Residue after evaporation ≤0.002 %
Free halogens Passes test
Titrable acid ≤0.0003 meq/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR