Benzoxazole 97+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,3-Benzoxazole

10568-0050 10568-0250 10568-1000
TS10568-0050EA 24 USD
TS10568-0050 TS10568-0250 TS10568-1000
Benzoxazole 97+%
Benzoxazole
Formula: C₇H₅NO
MW: 119.12 g/mol
Boiling Pt: 182…185 °C (1013 hPa)
Melting Pt: 27…30 °C
Flash Pt: 58 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005765
CAS Number: 273-53-0
EINECS: 205-988-9
UN: 1325
ADR: 4.1,III

Order Now

Specification Test Results

Appearance Colorless to white to yellow Low melting solid or liquid
Infrared spectrum Conforms
Melting point 27 to 30 °C
GC ≥97.0 %
UV at 280 nm A: ≤0.01
Titration after Ion exchange ≥98.5 % (On dry substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR