Benzofuran 99.5%

Supplier: Thermo Scientific Chemicals
Warning

10548-0100 10548-0500
TS10548-0100EA 79 USD
TS10548-0100 TS10548-0500
Benzofuran 99.5%
Benzofuran
Formula: C₈H₆O
Boiling Pt: 173…175 °C (760 mmHg)
Melting Pt: < –18 °C
Density: 1.07 g/cm³
Flash Pt: 50 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005847
CAS Number: 271-89-6
EINECS: 205-982-6
UN: 1993
ADR: 3,III
Merck Index: 13,01089

Order Now

Specification Test Results

Appearance Clear yellow to orange-brown Liquid
Infrared spectrum Conforms
Refractive index 1.5655 to 1.5685 (20°C, 589 nm)
GC ≥99.4 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR