7-Azaindole 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1H-Pyrrolo[2,3-b]pyridine

10510-0010 10510-0050 10510-0250
TS10510-0010EA 25 USD
TS10510-0010 TS10510-0050 TS10510-0250
7-Azaindole 98%
7-Azaindole
Formula: C₇H₆N₂
MW: 118.14 g/mol
Boiling Pt: 270 °C (1004HPA)
Melting Pt: 103…107 °C
Storage Temperature: Ambient
MDL Number: MFCD00005606
CAS Number: 271-63-6
EINECS: 205-981-0

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 103 to 107 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR