2-Aminopyrimidine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Pyrimidin-2-ylamine, Pyrimidin-2-amine

10458-0050 10458-1000 10458-5000
TS10458-0050EA 23 USD
TS10458-0050 TS10458-1000 TS10458-5000
2-Aminopyrimidine 98%
2-Aminopyrimidine
Formula: C₄H₅N₃
MW: 95.1 g/mol
Boiling Pt: 158 °C (186mbar)
Melting Pt: 124…127 °C
Flash Pt: 103 °C
Storage Temperature: Ambient
MDL Number: MFCD00006089
CAS Number: 109-12-6
EINECS: 203-648-4

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
Melting point 123 to 127 °C
Titration with HClO4 ≥97.5 %
BSC Certification Passes

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR