2-Aminopyrazine 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Aminopyrazine, Pyrazin-2-amine

10449-0100 10449-0500
TS10449-0100EA 88 USD
TS10449-0100 TS10449-0500
2-Aminopyrazine 99+%
2-Aminopyrazine
Formula: C₄H₅N₃
MW: 95.1 g/mol
Boiling Pt: 203 °C (1013 hPa)
Melting Pt: 117…120 °C
Storage Temperature: Ambient
MDL Number: MFCD00006137
CAS Number: 5049-61-6
EINECS: 225-748-7
UN: 1325
ADR: 4.1,III

Order Now

Specification Test Results

Appearance Light yellow to beige Crystalline powder
Infrared spectrum Conforms
Melting point 117 to 120 °C
Titration with HClO4 ≥99.0 %
Hydroxyl value ≥150 mg KOH/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR