2-Amino-6-methylpyridine 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 6-Methyl-2-pyridylamine, 6-Methylpyridin-2-amine, 2-Amino-6-picoline

10440-0050 10440-1000 10440-5000
TS10440-0050EA 25 USD
TS10440-0050 TS10440-1000 TS10440-5000
2-Amino-6-methylpyridine 98%
2-Amino-6-methylpyridine
Formula: C₆H₈N₂
MW: 108.14 g/mol
Boiling Pt: 208…209 °C (760 mmHg)
Melting Pt: 40…45 °C
Flash Pt: 103 °C
Storage Temperature: Ambient
MDL Number: MFCD00006331
CAS Number: 1824-81-3
EINECS: 217-360-1
UN: 2811
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Colorless to white to yellow to light orange Low melting crystalline solid
Infrared spectrum Conforms
Melting point 40 to 45 °C
GC ≥97.5 %
Particle size ≤0.4 % (dust)
pH 3.5 to 6

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR