2-Amino-5-methylpyridine 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 6-Amino-3-picoline, 5-Methyl-2-pyridylamine, 5-Methylpyridin-2-amine, 2-Amino-5-picoline

10439-0100 10439-0500
TS10439-0100EA 26 USD
TS10439-0100 TS10439-0500
2-Amino-5-methylpyridine 99%
2-Amino-5-methylpyridine
Formula: C₆H₈N₂
MW: 108.14 g/mol
Boiling Pt: 227.1 °C (1013 hPa)
Melting Pt: 80…81 °C
Storage Temperature: Ambient
MDL Number: MFCD00006328
CAS Number: 1603-41-4
EINECS: 216-503-5
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder or crystals or flakes
Infrared spectrum Conforms
Melting point 72 to 77 °C
Water ≤0.5 %
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR