2-Amino-3-methylpyridine 96%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-Methyl-2-pyridylamine, 3-Methylpyridin-2-amine, 2-Amino-3-picoline

10438-0050 10438-1000 10438-5000
TS10438-0050EA 27 USD
TS10438-0050 TS10438-1000 TS10438-5000
2-Amino-3-methylpyridine 96%
2-Amino-3-methylpyridine
Formula: C₆H₈N₂
MW: 108.14 g/mol
Boiling Pt: 221.1 °C (1013 hPa)
Melting Pt: 32…34 °C
Flash Pt: 111 °C
Storage Temperature: Ambient
MDL Number: MFCD00006320
CAS Number: 1603-40-3
EINECS: 216-501-4
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear yellow Liquid after melting
Infrared spectrum Conforms
Refractive index 1.5813 to 1.5833 (20°C, 589 nm)
Water ≤0.8 %
GC ≥95 %
Fluoride (F) ≤10 ppm
Arsenic (As) ≤3 ppm
Iron (Fe) ≤50 ppm
Lead (Pb) ≤5 ppm
Heavy metals (as Pb) ≤10 ppm
Insoluble matter ≤0.3 % (in water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR