6-Aminonicotinamide 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 6-Aminopyridine-3-carboxamide

10415-0010 10415-0050
TS10415-0010EA 85 USD
TS10415-0010 TS10415-0050
6-Aminonicotinamide 98%
6-Aminonicotinamide
Formula: C₆H₇N₃O
MW: 137.14 g/mol
Melting Pt: 245…248 °C
Storage Temperature: Ambient
MDL Number: MFCD00006327
CAS Number: 329-89-5

Order Now

Specification Test Results

Appearance White to light yellow to light grey Crystalline powder or crystals or solid
Infrared spectrum Conforms
Melting point 245 to 248 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR