2-(Aminomethyl)pyridine 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-Pyridylmethylamine, 2-Picolylamine, Pyridin-2-ylmethanamine, (2-Pyridyl)methylamine, 2-Picolamine
10407-0250
10407-1000
10407-5000
TS10407-0250EA
84
USD
TS10407-0250
TS10407-1000
TS10407-5000
2-(Aminomethyl)pyridine 99%
2-(Aminomethyl)pyridine
Formula:
C₆H₈N₂ Boiling Pt: 82…85 °C (12 mmHg) Melting Pt: –20 °C Density: 1.04 g/cm³ Flash Pt: 90 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006360 CAS Number: 3731-51-9 EINECS: 223-090-5 UN: 2735 ADR: 8,II |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5430 to 1.5470 (20°C, 589 nm) |
GC | ≥98.5 % |
Color scale | ≤15 APHA |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...