5-Aminoindole 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Indol-5-ylamine, 5-Indolamine, 5-Amino-1H-indole, 1H-Indol-5-amine

10390-0010 10390-0050
TS10390-0010EA 71 USD
TS10390-0010 TS10390-0050
5-Aminoindole 97%
5-Aminoindole
Formula: C₈H₈N₂
MW: 132.16 g/mol
Boiling Pt: 190 °C (6 mmHg)
Melting Pt: 130…134 °C
Storage Temperature: Freezer
MDL Number: MFCD00005679
CAS Number: 5192-03-0
EINECS: 225-977-2

Order Now

Specification Test Results

Appearance Off-white to grey-brown Crystalline powder
Infrared spectrum Conforms
Melting point 130 to 134 °C
HPLC ≥96.0 %
Titration with HClO4 ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR