1-(2-Aminoethyl)pyrrolidine 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N-(2-Aminoethyl)pyrrolidine, Pyrrolidinoethylamine

10370-0050 10370-0250 10370-1000
TS10370-0050EA 69 USD
TS10370-0050 TS10370-0250 TS10370-1000
1-(2-Aminoethyl)pyrrolidine 99%
1-(2-Aminoethyl)pyrrolidine
Formula: C₆H₁₄N₂
MW: 114.19 g/mol
Boiling Pt: 66…70 °C (23 mmHg)
Density: 0.9 g/cm³
Flash Pt: 47 °C
Storage Temperature: Ambient
MDL Number: MFCD00003182
CAS Number: 7154-73-6
EINECS: 230-509-5
UN: 1993
ADR: 3 III

Order Now

Specification Test Results

Appearance Clear colorless to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.4677 to 1.4697 (20°C, 589 nm)
GC ≥88 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR