2-Amino-4,6-dihydroxypyrimidine 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 2-Amino-6-hydroxy-1H-pyrimidin-4-one, 2-Amino-6-hydroxypyrimidin-4(3H)-one

10351-0250 10351-1000
TS10351-0250EA 68 USD
TS10351-0250 TS10351-1000
2-Amino-4,6-dihydroxypyrimidine 98%
2-Amino-4,6-dihydroxypyrimidine
Formula: C₄H₅N₃O₂
MW: 127.1 g/mol
Boiling Pt: 394 °C (1013 hPa)
Melting Pt: >300 °C
MDL Number: MFCD00006094
CAS Number: 56-09-7
EINECS: 200-256-5

Order Now

Specification Test Results

Appearance Off-white to light yellow to light pink Powder
Infrared spectrum Conforms
Titration excess NaOH ≥97.5 %
UV Lambda max 628 to 632 nm
Absorption Ratio P-15/P+15 0.94 to 1.14
BSC Certification Passes

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR