3-Amino-2-chloropyridine 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Chloro-3-pyridylamine, 2-Chloropyridin-3-amine

10342-0100 10342-0500
TS10342-0100EA 39 USD
TS10342-0100 TS10342-0500
3-Amino-2-chloropyridine 96%
3-Amino-2-chloropyridine
Formula: C₅H₅ClN₂
MW: 128.56 g/mol
Boiling Pt: 263 °C (1013 hPa)
Melting Pt: 77…82 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006238
CAS Number: 6298-19-7
EINECS: 228-572-9

Order Now

Specification Test Results

Appearance Off-white to yellow to pink Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 76 to 81 °C
HPLC ≥95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR