4-Amino-6-chloro-2-(methylthio)pyrimidine 97%

Supplier: Thermo Scientific Chemicals
Warning

10339-0010 10339-0050 10339-0250
TS10339-0010EA 37 USD
TS10339-0010 TS10339-0050 TS10339-0250
4-Amino-6-chloro-2-(methylthio)pyrimidine 97%
4-Amino-6-chloro-2-(methylthio)pyrimidine
Formula: C₅H₆ClN₃S
MW: 175.6400 g/mol
Melting Pt: 130…134 °C
Storage Temperature: Ambient
MDL Number: MFCD00006088
CAS Number: 1005-38-5

Order Now

Specification Test Results

Appearance Off-white to yellow to beige Crystalline powder
Infrared spectrum Conforms
Melting point 130 to 134 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR