α-Acetyl-γ-butyrolactone 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: alpha-Acetyl-gamma-hydroxybutyric acid Lactone, α-Acetyl-γ-hydroxybutyric acid Lactone, 3-Acetyldihydrofuran-2(3H)-one, 2-Acetyl-γ-butyrolactone, 2-Acetylbutyrolactone, alpha-Acetyl-gamma-butyrolactone, α-Acetylbutyrolactone, 3-Acetyl-4,5-dihydro-2(3H)-furanone

10249-0010 10249-1000 10249-2500
TS10249-0010EA 195 USD
TS10249-0010 TS10249-1000 TS10249-2500
α-Acetyl-γ-butyrolactone 99+%
α-Acetyl-γ-butyrolactone
Formula: C₆H₈O₃
Boiling Pt: 107…108 °C (5 mmHg)
Density: 1.18 g/cm³
Flash Pt: 140 °C
Storage Temperature: Ambient
MDL Number: MFCD00005394
CAS Number: 517-23-7
EINECS: 208-235-2

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4580 to 1.4610 (20°C, 589 nm)
GC ≥99.0 %
Color scale ≤30 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR