4-(4-Morpholinylsulfonyl)benzeneboronic acid
Supplier: Matrix Scientific
Synonyms:
4-(Morpholinosulphonyl)phenylboronic acid, 4-Boronophenyl morpholin-4-yl sulphone, 4-[(Morpholin-4-yl)sulphonyl]benzeneboronic acid, 4-[(4-Boronophenyl)sulphonyl]morpholine
148199-1G
148199-5G
103653-042EA
307.69
USD
103653-042
103653-044
4-(4-Morpholinylsulfonyl)benzeneboronic acid
4-(4-Morpholinylsulfonyl)benzeneboronic acid
Formula:
C₁₀H₁₄BNO₅S MW: 271.09 g/mol |
MDL Number:
MFCD06659844 CAS Number: 486422-68-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...