Methyl 2-(benzylamino)acetate hydrochloride ≥99% (by HPLC)

Supplier: CHEM IMPEX INTL INC MS

Synonyms: Bzl-Gly-OMe·HCl, Benzyl glycine methyl ester hydrochloride, N-Benzylglycine methyl ester hydrochloride

11912-1G 11912-25G 11912-5G
102593-796EA 53.57 USD
102593-796 102593-798 102639-356
Methyl 2-(benzylamino)acetate hydrochloride ≥99% (by HPLC)
Methyl 2-(benzylamino)acetate hydrochloride
Formula: C₁₀H₁₃NO₂.HCl
MW: 215.68 g/mol
Melting Pt: 133…140 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02684379
CAS Number: 17136-35-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR