Pyrrolo[2,1-f][1,2,4]triazin-4-amine

Supplier: Thermo Scientific Chemicals
Warning

46899-0010 468990050
TS46899-0010EA 59 USD
TS46899-0010 TS468990050
Pyrrolo[2,1-f][1,2,4]triazin-4-amine
Pyrrolo[2,1-f][1,2,4]triazin-4-amine
Formula: C₆H₆N₄
MW: 134.14 g/mol
MDL Number: MFCD08234647
CAS Number: 159326-68-8

Order Now

Specification Test Results

HPLC ≥94 %
Appearance (Color) Almost white to grey
Appearance (Form) Powder
Infrared spectrum Conforms


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR