4-Methyltetrahydropyran 95% stabilized

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-Methyltetrahydro-2H-pyran, 4-Methyloxane

Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)

46732-0010 46732-1000 467320025
TS46732-0010EA 50 USD
TS46732-0010 TS46732-1000 TS467320025
4-Methyltetrahydropyran 95% stabilized
4-Methyltetrahydropyran
Formula: C₆H₁₂O
MW: 100.16 g/mol
Boiling Pt: 105 °C
MDL Number: MFCD07783308
CAS Number: 4717-96-8
EINECS: 225-207-5
UN: 2924
ADR: 3.8,II

Order Now

Specification Test Results

Appearance (Color) Clear colorless
Color ≤10 APHA
Refractive index 1.4170 to 1.4200 (20°C, 589 nm)
Appearance (Form) liquid
Infrared spectrum Conforms
GC ≥98.0 %
Stabilizer 20 ppm BHT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR