3-Hydroxyazetidine 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Azetidinol, Azetidin-3-ol

46721-0010 467210050
TS46721-0010EA 71 USD
TS46721-0010 TS467210050
3-Hydroxyazetidine 95%
3-Hydroxyazetidine
Formula: C₃H₇NO
MW: 73.09 g/mol
MDL Number: MFCD03695446
CAS Number: 45347-82-8

Order Now

Specification Test Results

Infrared spectrum Conforms
Appearance (Color) White to grey
Appearance (Form) Powder
Titration with HClO4 ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR