Pyrroloquinoline quinone 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4,5-Dioxo-4,5-dihydro-1H-pyrrolo[2,3-f]quinoline-2,7,9-tricarboxylic acid, 4,5-Dihydro-4,5-dioxo-1H-pyrrolo[2,3-f]quinoline-2,7,9-tricarboxylic acid, PQQ, Methoxatin

46701-0010 467012500
TS46701-0010EA 819 USD
TS46701-0010 TS467012500
Pyrroloquinoline quinone 95%
Pyrroloquinoline quinone
Formula: C₁₄H₆N₂O₈
MW: 330.21 g/mol
MDL Number: MFCD00043125
CAS Number: 72909-34-3

Order Now

Specification Test Results

Appearance (Color) Dark orange to red to brown
Appearance (Form) powder
Infrared spectrum Conforms
HPLC ≥95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR