Azetidine-2-carboxylic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
46690-0050
466900010
TS46690-0050EA
349
USD
TS46690-0050
TS466900010
Azetidine-2-carboxylic acid 97%
Formula:
C₄H₇NO₂ |
MDL Number:
MFCD00066660 CAS Number: 2517-04-6 |
Specification Test Results
Appearance (Color) | White to almost white |
Appearance (Form) | Crystalline powder |
Infrared spectrum | Conforms |
Water | ≤1 % |
Titration with HClO4 | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...