1-(tert-Butoxycarbonyl)-2-pyrrolidinone 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(tert-Butoxycarbonyl)-2-oxopyrrolidine, tert-Butyl 2-oxopyrrolidine-1-carboxylate, Pyrrolidin-2-one, N-BOC protected

46633-0250 466330050
TS46633-0250EA 140 USD
TS46633-0250 TS466330050
1-(tert-Butoxycarbonyl)-2-pyrrolidinone 97%
1-(tert-Butoxycarbonyl)-2-pyrrolidinone
Formula: C₉H₁₅NO₃
MW: 185.22 g/mol
Density: 1.086 g/mL (25 °C) (lit.)
MDL Number: MFCD00674099
CAS Number: 85909-08-6

Order Now

Specification Test Results

Appearance (Color) Clear colorless to yellow to brown
Appearance (Form) Liquid
Infrared spectrum Conforms
Refractive index 1.4645 to 1.4670 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR