Atropic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Phenylacrylic Acid, Ipratropium bromide Impurity D

45905-0250 45905-0050
TS45905-0250EA 203 USD
TS45905-0250 TS45905-0050
Atropic acid 97%
Atropic acid
Formula: C₉H₈O₂
MW: 148.16 g/mol
Boiling Pt: 267 °C
Melting Pt: 104…106 °C
Storage Temperature: Ambient
MDL Number: MFCD00046531
CAS Number: 492-38-6

Order Now

Specification Test Results

Appearance White to light yellow Powder or crystalline powder
Infrared spectrum Conforms
Melting point 104 to 110 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR