Tripropylene glycol (mixture of isomers) 99%, pure
Supplier: Thermo Scientific Chemicals
Synonyms:
Tripropylene glycol
45848-0010
45848-0050
45848-0250
TS45848-0010EA
38
USD
TS45848-0010
TS45848-0050
TS45848-0250
Tripropylene glycol (mixture of isomers) 99%, pure
Tripropylene glycol (mixture of isomers)
Formula:
C₉H₂₀O₄ MW: 192.26 g/mol Boiling Pt: 270 °C (760 mmHg) Density: 1.02 g/cm³ Flash Pt: 145 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014405 CAS Number: 24800-44-0 |
Specification Test Results
Appearance | Clear colorless Viscous liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4430 to 1.4450 (20°C, 589 nm) |
Water | ≤0.05 % |
GC | ≥98.5 % (sum of isomers) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...