5-Bromo-1,2,3-trimethoxybenzene 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Bromo-3,4,5-trimethoxybenzene

45813-0250 45813-1000 45813-0050
TS45813-0250EA 130 USD
TS45813-0250 TS45813-1000 TS45813-0050
5-Bromo-1,2,3-trimethoxybenzene 97%
5-Bromo-1,2,3-trimethoxybenzene
Formula: C₉H₁₁BrO₃
Melting Pt: 78…82 °C
Storage Temperature: Ambient
MDL Number: MFCD00017169
CAS Number: 2675-79-8

Order Now

Specification Test Results

Appearance White to brown Powder or crystalline powder
Infrared spectrum Conforms
Melting point 75 to 83 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR