(2-Fluoropyridin-4-yl)methanol 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (2-fluoro-pyridin-4-yl)-methanol, 2-Fluoro-4-hydroxymethylpyridine

45237-0010 45237-0050
TS45237-0010EA 153 USD
TS45237-0010 TS45237-0050
(2-Fluoropyridin-4-yl)methanol 97%
(2-Fluoropyridin-4-yl)methanol
Formula: C₆H₆FNO
MW: 127.12 g/mol
Melting Pt: 61…63 °C
Storage Temperature: Ambient
MDL Number: MFCD06200782
CAS Number: 131747-60-9

Order Now

Specification Test Results

Appearance White to off-white to pink Powder
Infrared spectrum Conforms
Melting point 61 to 67 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR