3-Methylstyrene 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-Vinyltoluene

45087-0010 45087-0050
TS45087-0010EA 40 USD
TS45087-0010 TS45087-0050
3-Methylstyrene 95%
3-Methylstyrene
Formula: C₉H₁₀
MW: 118.18 g/mol
Boiling Pt: 170…171 °C
Melting Pt: 82…81 °C
Flash Pt: 51 °C
Storage Temperature: Ambient
MDL Number: MFCD00008617
CAS Number: 100-80-1
UN: 2618
ADR: 3,III

Order Now

Specification Test Results

Appearance Colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥94.0 %
Stabilizer 0.1 % 4-tert-Butylcatechol

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR