Atenolol 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 2-[4-(2-Hydroxy-3-isopropylaminopropoxy)phenyl]acetamide

44955-0010 44955-0050 44955-0250
TS44955-0010EA 62 USD
TS44955-0010 TS44955-0050 TS44955-0250
Atenolol 98%
Atenolol
Formula: C₁₄H₂₂N₂O₃
MW: 266.34 g/mol
Melting Pt: 146…148 °C
Storage Temperature: Ambient
MDL Number: MFCD00057645
CAS Number: 29122-68-7

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 151 to 155 °C
Titration with HClO4 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR