4-Amino-2-(trifluoromethyl)benzonitrile 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Cyano-3-trifluoromethylaniline

44247-0050 44247-0010
TS44247-0050EA 253 USD
TS44247-0050 TS44247-0010
4-Amino-2-(trifluoromethyl)benzonitrile 97%
4-Amino-2-(trifluoromethyl)benzonitrile
Formula: C₈H₅F₃N₂
MW: 186.14 g/mol
Melting Pt: 144…146 °C
Storage Temperature: Ambient
MDL Number: MFCD00042155
CAS Number: 654-70-6
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to orange to brown Powder or crystals
Infrared spectrum Conforms
Melting point 138 to 146 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR