4,5-Dimethoxy-2-nitrobenzyl bromide 97%

Supplier: Thermo Scientific Chemicals
Danger

44077-0010 44077-0050
TS44077-0010EA 120 USD
TS44077-0010 TS44077-0050
4,5-Dimethoxy-2-nitrobenzyl bromide 97%
4,5-Dimethoxy-2-nitrobenzyl bromide
Formula: C₉H₁₀BrNO₄
MW: 276.086 g/mol
Melting Pt: 130…132 °C
Storage Temperature: Ambient
MDL Number: MFCD00192064
CAS Number: 53413-67-5
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance Yellow to brown Crystalline powder or needles
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR