2-Chlorophenethyl bromide 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1-(2-Bromoethyl)-2-chlorobenzene, 2-Chlorophenethyl Bromide, 2-(2-Chlorophenyl)ethyl bromide, 2-Chlorophenethyl bromide
44014-0010
44014-0050
TS44014-0010EA
41
USD
TS44014-0010
TS44014-0050
2-Chlorophenethyl bromide 95%
2-Chlorophenethyl bromide
Formula:
C₈H₈BrCl MW: 219.51 g/mol Boiling Pt: 110…112 °C (10Torr) Density: 1.4569 g/cm³ Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD04117464 CAS Number: 16793-91-2 UN: 3082 ADR: 9 III |
Specification Test Results
Appearance | Colorless to yellow Liquid |
Infrared spectrum | Conforms |
GC | ≥94.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...