2-Fluorophenethyl bromide 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-(2-Bromoethyl)-2-fluorobenzene
44012-0050
44012-0010
TS44012-0050EA
202
USD
TS44012-0050
TS44012-0010
2-Fluorophenethyl bromide 97%
2-Fluorophenethyl bromide
Formula:
C₈H₈BrF MW: 203.05 g/mol Boiling Pt: 73…75 °C (20Torr) Storage Temperature: Ambient |
MDL Number:
MFCD04117466 CAS Number: 91319-54-9 UN: 3082 ADR: 9 III |
Specification Test Results
Appearance | Colorless to yellow Liquid |
Infrared spectrum | Conforms |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...