4-(Chloromethyl)benzyl alcohol 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: p-hydroxymethylbenzyl chloride, (4-(Chloromethyl)phenyl)methanol, 4-(hydroxymethyl)benzyl chloride, Benzenemethanol,4-(chloromethyl)-

43892-0010 43892-0050
TS43892-0010EA 56 USD
TS43892-0010 TS43892-0050
4-(Chloromethyl)benzyl alcohol 97%
4-(Chloromethyl)benzyl alcohol
Formula: C₈H₉ClO
MW: 156.609 g/mol
Melting Pt: 58…60 °C
Storage Temperature: Ambient
MDL Number: MFCD03427011
CAS Number: 16473-35-1
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder or solid
Infrared spectrum Conforms
Melting point 57 to 61 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR