3-(Dimethylamino)benzyl alcohol 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (3-(Dimethylamino)phenyl)methanol, m-(Dimethylamino)benzyl alcohol, Benzenemethanol,3-(dimethylamino)-, Benzyl alcohol, m-(dimethylamino)-

43816-0010 43816-0050
TS43816-0010EA 54 USD
TS43816-0010 TS43816-0050
3-(Dimethylamino)benzyl alcohol 97%
3-(Dimethylamino)benzyl alcohol
Formula: C₉H₁₃NO
Boiling Pt: 109…110 °C (.1Torr)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00017342
CAS Number: 23501-93-1

Order Now

Specification Test Results

Appearance Clear colorless to yellow to light brown Liquid
Infrared spectrum Conforms
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR