3-Amino-1-adamantanol 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Aminoadamantan-1-ol

43538-0010 43538-0050
TS43538-0010EA 65 USD
TS43538-0010 TS43538-0050
3-Amino-1-adamantanol 96%
3-Amino-1-adamantanol
Formula: C₁₀H₁₇NO
MW: 167.25 g/mol
Melting Pt: 263…264 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD01821204
CAS Number: 702-82-9

Order Now

Specification Test Results

Appearance White to yellow Crystalline powder
Infrared spectrum Conforms
Water ≤10 % (K.F.)
Elemental Analysis 7.55 to 8.35 % N

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR