1-Octanol, anhydrous 99%, AcroSeal™

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: n-Octanol

43458-0010 43458-1000
TS43458-0010EA 167 USD
TS43458-0010 TS43458-1000
1-Octanol, anhydrous 99%, AcroSeal™
1-Octanol
Formula: H₃C(CH₂)₆CH₂OH
MW: 130.23 g/mol
Boiling Pt: 195 °C (760 mmHg)
Melting Pt: –16 °C
Density: 0.824 g/cm³
Flash Pt: 81 °C
Storage Temperature: Ambient
MDL Number: MFCD00002988
CAS Number: 111-87-5
EINECS: 203-917-6
Merck Index: 13,06784
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7db4a6be3aedee4af43781ec6deb31631bc376c1&vid=80cb13f618a2d3181daf8310324993f1f7c69fb2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.005 %
GC ≥98.5 %
Residue after evaporation ≤0.004 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR