4-(tert-Butoxycarbonylamino)-1-butanol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
tert-Butyl (4-hydroxybutyl)carbamate, N-Boc-4-aminobutanol, 4-(Boc-amino)-1-butanol, N-(4-Hydroxybutyl)carbamic Acid tert-Butyl Ester, tert-Butyl N-(4-Hydroxybutyl)carbamate
43421-0010
43421-0050
TS43421-0010EA
81
USD
TS43421-0010
TS43421-0050
4-(tert-Butoxycarbonylamino)-1-butanol 98%
4-(tert-Butoxycarbonylamino)-1-butanol
Formula:
C₉H₁₉NO₃ MW: 189.26 g/mol Melting Pt: 37…40 °C Density: 1.02 g/cm³ Storage Temperature: Refrigerator |
MDL Number:
MFCD01861334 CAS Number: 75178-87-9 |
Specification Test Results
Appearance | Colorless to yellow Liquid or low melting solid |
Infrared spectrum | Conforms |
GC | ≥97.5 % |
HPLC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...