1-Hexanol, anhydrous 99%, AcroSeal™
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Hexyl alcohol, Pentyl carbinol, 1-Hexyl alcohol, 1-Hydroxy hexane
43386-0010
43386-1000
TS43386-0010EA
183
USD
TS43386-0010
TS43386-1000
1-Hexanol, anhydrous 99%, AcroSeal™
1-Hexanol
Formula:
H₃C(CH₂)₄CH₂OH MW: 102.18 g/mol Boiling Pt: 156…157 °C (760 mmHg) Melting Pt: –52 °C Density: 0.8136 g/cm³ Flash Pt: 59 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002982 CAS Number: 111-27-3 UN: 2282 ADR: 3,III |
Specification Test Results
Appearance | Clear colorless Liquid |
Water | ≤0.005 % |
GC | ≥98.5 % |
Residue after evaporation | ≤0.002 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...