Thiophenol 99%, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

43072-1000 43072-8000
TS43072-1000EA 53 USD
TS43072-1000 TS43072-8000
Thiophenol 99%, AcroSeal™
Thiophenol
Formula: C₆H₆S
Boiling Pt: 167…169 °C (760 mmHg)
Melting Pt: –15 °C
Density: 1.078 g/cm³
Flash Pt: 50 °C
Storage Temperature: Ambient
MDL Number: MFCD00004826
CAS Number: 108-98-5
UN: 2337
ADR: 6.1,I

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5880 to 1.5900 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR