2-Formylbenzeneboronic acid pinacol ester 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Formylphenylboronic acid Pinacol ester, 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde, 2-Formylphenylboronic acid pinacol ester, 2-(2-Formylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

43067-0050 43067-0010
TS43067-0050EA 154 USD
TS43067-0050 TS43067-0010
2-Formylbenzeneboronic acid pinacol ester 97%
2-Formylbenzeneboronic acid pinacol ester
Formula: C₁₃H₁₇BO₃
Storage Temperature: Refrigerator
MDL Number: MFCD07363841
CAS Number: 380151-85-9

Order Now

Specification Test Results

Appearance Clear colorless to dark yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR