4-Fluoro-2-(trifluoromethyl)phenylboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Fluoro-2-(trifluoromethyl)benzeneboronic acid, (4-Fluoro-2-(trifluoromethyl)phenyl)boronic acid

43044-0010 43044-0050
TS43044-0010EA 115 USD
TS43044-0010 TS43044-0050
4-Fluoro-2-(trifluoromethyl)phenylboronic acid 97%
4-Fluoro-2-(trifluoromethyl)phenylboronic acid
Formula: C₇H₅BF₄O₂
MW: 207.92 g/mol
Melting Pt: 184…186 °C
Storage Temperature: Ambient
MDL Number: MFCD08062379
CAS Number: 182344-16-7

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Melting point 182 to 187 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR