2-(Propyloxy)ethanol 98%, pure
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Ethylene glycol monopropyl ether, 2-Propoxyethanol, Propyl glycol
41840-0010
41840-0050
TS41840-0010EA
64
USD
TS41840-0010
TS41840-0050
2-(Propyloxy)ethanol 98%, pure
2-(Propyloxy)ethanol
Formula:
C₅H₁₂O₂ MW: 104.15 g/mol Melting Pt: –70 °C Flash Pt: 57 °C Storage Temperature: Ambient |
MDL Number:
MFCD00072645 CAS Number: 2807-30-9 EINECS: 220-548-6 UN: 1986 ADR: 3 III |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4120 to 1.4140 (20°C, 589 nm) |
Water | ≤0.1 % (K.F.) |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...