Biphenyl-3-ol 90%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Phenylphenol, [1,1'-Biphenyl]-3-ol, 3-Hydroxybiphenyl, 3-Hydroxydiphenyl

41766-0050 41766-0250
TS41766-0050EA 77 USD
TS41766-0050 TS41766-0250
Biphenyl-3-ol 90%
Biphenyl-3-ol
Formula: C₁₂H₁₀O
MW: 170.21 g/mol
Boiling Pt: 300 °C (760 mmHg)
Melting Pt: 72…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00002294
CAS Number: 580-51-8

Order Now

Specification Test Results

Appearance White to yellow to pink to light brown Crystalline powder and/or chunks
Infrared spectrum Conforms
Melting point 72 to 80 °C
GC ≥88 %
Isomer balance mainly para-isomer

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR