5-Nitrosalicylaldehyde 98+%

Supplier: Thermo Scientific Chemicals
Warning

41618-0050 41618-0250
TS41618-0050EA 45 USD
TS41618-0050 TS41618-0250
5-Nitrosalicylaldehyde 98+%
5-Nitrosalicylaldehyde
Formula: C₇H₅NO₄
Melting Pt: 125…129 °C
Storage Temperature: Ambient
MDL Number: MFCD00007337
CAS Number: 97-51-8
EINECS: 202-587-0

Order Now

Specification Test Results

Appearance Yellow to orange to brown Powder or crystals
Infrared spectrum Conforms
Melting point 124 to 128 °C
GC ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR